Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pinx1
Entrez Gene ID
448337
Gene Name
PIN2 (TERF1) interacting telomerase inhibitor 1
Gene Aliases
Gno1, Pxr1, lptl, lpts
Chromosome Location
Chr.Un|NW_016683369: 289679 - 361559 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004938.1 NP_001004938.1 4 - 5 350 84
GenBank mRNA BC075416.1 - 4 - 5 350 84
EU520259.1 - 4 - 5 297 84

View Full Product Details