Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pinx1
Entrez Gene ID
448337
Gene Name
PIN2 (TERF1) interacting telomerase inhibitor 1
Gene Aliases
Gno1, Pxr1, lptl, lpts
Chromosome Location
Chr.Un|NW_016683369: 289679 - 361559 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004938.1 NP_001004938.1 5 - 6 441 81
XM_012964033.2 XP_012819487.1 5 - 6 487 81
GenBank mRNA BC075416.1 - 5 - 6 441 81
CR761068.2 - 5 - 6 416 81
CT485754.1 - - 485 81
EU520259.1 - 5 - 6 388 81

View Full Product Details