Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ovol2
Entrez Gene ID
448478
Gene Name
ovo like zinc finger 2
Gene Aliases
znf339
Chromosome Location
Chr.5: 38586426 - 38618234 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004998.1 NP_001004998.1 1 - 2 315 84
GenBank mRNA BC075593.1 - 1 - 2 315 84
CR855435.2 - 1 - 2 391 84

View Full Product Details