Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ovol2
Entrez Gene ID
448478
Gene Name
ovo like zinc finger 2
Gene Aliases
znf339
Chromosome Location
Chr.5: 38586426 - 38618234 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004998.1 NP_001004998.1 4 - 4 2190 52
GenBank mRNA BC075593.1 - 4 - 4 2190 52

View Full Product Details