Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
c19orf47
Entrez Gene ID
448521
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.8: 56784568 - 56795053 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005016.1 NP_001005016.1 4 - 5 342 96
GenBank mRNA BC076686.1 - 4 - 5 342 96
CR848118.2 - 4 - 5 287 96

View Full Product Details