The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005024.1 | NP_001005024.1 | 2 - 3 | 255 | 89 |
GenBank mRNA | BC076705.1 | - | 2 - 3 | 255 | 89 |
CR760251.2 | - | 2 - 3 | 190 | 89 |