Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chmp1b
Entrez Gene ID
448582
Gene Name
charged multivesicular body protein 1B
Gene Aliases
-
Chromosome Location
Chr.8: 43234604 - 43243286 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005047.1 NP_001005047.1 4 - 5 409 99
GenBank mRNA BC076916.1 - 4 - 5 409 99
CR760798.2 - 4 - 5 391 99

View Full Product Details