Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fuca2
Entrez Gene ID
496744
Gene Name
alpha-L-fucosidase 2
Gene Aliases
-
Chromosome Location
Chr.5: 67422242 - 67459679 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC088008.1 - 4 - 5 1024 79

View Full Product Details