Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pnhd
Entrez Gene ID
394459
Gene Name
pinhead
Gene Aliases
-
Chromosome Location
Chr.1: 85454509 - 85462203 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203534.1 NP_988865.1 3 - 4 346 87
GenBank mRNA AY333424.1 - 3 - 4 346 87
BC170803.1 - 3 - 4 337 87
BC170805.1 - 3 - 4 337 87
CR926370.2 - 3 - 4 322 87

View Full Product Details