Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ing5
Entrez Gene ID
394462
Gene Name
inhibitor of growth family member 5
Gene Aliases
p28ing5
Chromosome Location
Chr.5: 122141482 - 122158683 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203536.1 NP_988867.1 1 - 2 99 80
GenBank mRNA BC059768.1 - 1 - 2 99 80
CT030479.1 - 1 - 2 76 80

View Full Product Details