Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myl1
Entrez Gene ID
394551
Gene Name
myosin light chain 1
Gene Aliases
MLC1f/3f, XMLC1f, XMLC3f, mlc1f, mlc3f
Chromosome Location
Chr.9: 51701081 - 51714255 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203623.1 NP_988954.1 4 - 5 588 78
XM_012970239.2 XP_012825693.1 4 - 5 711 78
GenBank mRNA BC061302.1 - 4 - 5 588 78
BC089727.1 - 4 - 5 574 78
BC169133.1 - - 571 78
CR760864.2 - 4 - 5 557 78
CR926445.2 - 4 - 5 387 78

View Full Product Details