Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2m
Entrez Gene ID
394553
Gene Name
ubiquitin conjugating enzyme E2M
Gene Aliases
-
Chromosome Location
Chr.7: 94883298 - 94893363 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203625.1 NP_988956.1 1 - 2 247 83
GenBank mRNA BC061289.1 - 1 - 2 247 83

View Full Product Details