Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
elob
Entrez Gene ID
394559
Gene Name
elongin B
Gene Aliases
elongin-B, elonginb, siii, tceb2
Chromosome Location
Chr.9: 7142775 - 7150904 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203631.1 NP_988962.1 2 - 3 191 78
GenBank mRNA BC061439.1 - 2 - 3 191 78
BC084191.1 - 2 - 3 221 78

View Full Product Details