Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmb1
Entrez Gene ID
394589
Gene Name
proteasome subunit beta 1
Gene Aliases
-
Chromosome Location
Chr.5: 40044868 - 40050248 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203662.1 NP_988993.1 4 - 5 493 70
GenBank mRNA BC061284.1 - 4 - 5 493 70
CR760776.2 - 4 - 5 498 70

View Full Product Details