Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp5pb
Entrez Gene ID
394723
Gene Name
ATP synthase peripheral stalk-membrane subunit b
Gene Aliases
Fo-b, atp5f1, pig47
Chromosome Location
Chr.2: 40271863 - 40280519 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203787.1 NP_989118.1 6 - 7 710 108
GenBank mRNA BC061296.1 - 6 - 7 710 108
CR760878.2 - 6 - 7 742 108

View Full Product Details