Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hmces
Entrez Gene ID
394754
Gene Name
5-hydroxymethylcytosine binding, ES cell specific
Gene Aliases
c3orf37
Chromosome Location
Chr.4: 125147459 - 125157174 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203818.1 NP_989149.1 4 - 5 503 92
XM_012960759.2 XP_012816213.1 4 - 5 569 92
GenBank mRNA BC061596.1 - 4 - 5 503 92
CR761600.2 - 4 - 5 501 92

View Full Product Details