Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sec22b
Entrez Gene ID
394761
Gene Name
uncharacterized LOC394761
Gene Aliases
ers-24, sec22b-a, sec22b-b, sec22l1
Chromosome Location
Chr.4: 107712846 - 107721621 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203825.1 NP_989156.1 4 - 5 511 88
GenBank mRNA BC061616.1 - 4 - 5 511 88

View Full Product Details