Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
calhm2
Entrez Gene ID
394768
Gene Name
calcium homeostasis modulator 2
Gene Aliases
fam26b
Chromosome Location
Chr.7: 20315201 - 20330682 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203830.1 NP_989161.1 2 - 3 638 100
GenBank mRNA BC063213.1 - 2 - 3 638 100

View Full Product Details