Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alg11
Entrez Gene ID
394898
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Gene Aliases
gt8, utp14c
Chromosome Location
Chr.2: 147180123 - 147187689 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203952.1 NP_989283.1 1 - 2 37 82
GenBank mRNA BC064222.1 - 1 - 2 37 82

View Full Product Details