Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcat
Entrez Gene ID
394899
Gene Name
glycine C-acetyltransferase
Gene Aliases
kbl
Chromosome Location
Chr.4: 112127888 - 112145074 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203953.1 NP_989284.1 7 - 8 1032 92
GenBank mRNA BC064221.1 - 7 - 8 1032 92
CR761814.2 - 7 - 8 1014 92

View Full Product Details