Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cndp2
Entrez Gene ID
394907
Gene Name
CNDP dipeptidase 2 (metallopeptidase M20 family)
Gene Aliases
cn2, cpgl, darmin-r
Chromosome Location
Chr.Un|NW_016683375: 1670074 - 1684505 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203958.1 NP_989289.1 1 - 2 138 93
GenBank mRNA BC064175.1 - 1 - 2 138 93

View Full Product Details