Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cnih4
Entrez Gene ID
394910
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Gene Aliases
CNIH-4
Chromosome Location
Chr.5: 5079372 - 5088507 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203961.1 NP_989292.1 4 - 5 417 91
XM_012962850.1 XP_012818304.1 3 - 4 1194 91
GenBank mRNA BC063923.1 - 4 - 5 417 91

View Full Product Details