Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnajc9
Entrez Gene ID
394914
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
-
Chromosome Location
Chr.7: 32263556 - 32271561 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203965.1 NP_989296.1 4 - 5 672 94
GenBank mRNA BC064229.1 - 4 - 5 672 94

View Full Product Details