Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tagln2
Entrez Gene ID
394981
Gene Name
transgelin 2
Gene Aliases
sm22-a, sm22-alpha
Chromosome Location
Chr.8: 115178329 - 115209863 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204023.1 NP_989354.1 3 - 4 392 74
XM_012969825.1 XP_012825279.1 3 - 4 508 74
XM_012969826.1 XP_012825280.1 3 - 4 427 74
GenBank mRNA BC064157.1 - 3 - 4 392 74
CT025168.2 - 2 - 3 382 74

View Full Product Details