Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp3r1
Entrez Gene ID
395037
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Gene Aliases
-
Chromosome Location
Chr.5: 30906249 - 30930146 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204069.1 NP_989400.1 5 - 6 621 133
GenBank mRNA AY336972.1 - 5 - 6 622 133
BC064854.1 - 5 - 6 621 133

View Full Product Details