Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dtx2
Entrez Gene ID
407940
Gene Name
deltex 2, E3 ubiquitin ligase
Gene Aliases
-
Chromosome Location
Chr.2: 90292445 - 90342768 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213669.2 NP_998834.1 8 - 9 1702 93
XM_012956690.2 XP_012812144.1 8 - 9 1737 93
XM_012956691.2 XP_012812145.1 8 - 9 1676 93
XM_018091155.1 XP_017946644.1 7 - 8 1535 93
GenBank mRNA BC067930.2 - 8 - 9 1702 93

View Full Product Details