Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ag1
Entrez Gene ID
407906
Gene Name
anterior gradient 1
Gene Aliases
XAG, Xag2, ag, ag1-a, ag1-b, xag-1, xag1
Chromosome Location
Chr.9: 55625443 - 55632154 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213699.1 NP_998864.1 6 - 7 507 61
GenBank mRNA BC067924.1 - 6 - 7 507 61
CT030434.1 - 6 - 7 469 61

View Full Product Details