Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
slc25a22
Entrez Gene ID
447993
Gene Name
solute carrier family 25 (mitochondrial carrier: glutamate), member 22
Gene Aliases
slc25a18
Chromosome Location
Chr.2: 66717630 - 66735896 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005430.1 NP_001005430.1 7 - 8 621 101
XM_012958836.2 XP_012814290.1 4 - 5 3442 101
GenBank mRNA BC074507.1 - 7 - 8 621 101

View Full Product Details