Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab27b
Entrez Gene ID
448182
Gene Name
RAB27B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 190001592 - 190129826 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005681.2 NP_001005681.1 2 - 3 226 84
NM_001195546.1 NP_001182475.1 4 - 5 493 84
GenBank mRNA BC075102.1 - 2 - 3 226 84
DN070478.1 - 3 - 4 320 84
DN097288.1 - 4 - 5 493 84

View Full Product Details