Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccni
Entrez Gene ID
448195
Gene Name
cyclin I
Gene Aliases
ccni-a, ccni-b, cyc1, cyi
Chromosome Location
Chr.1: 94785054 - 94863001 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005690.1 NP_001005690.1 4 - 5 514 71
XM_012959253.1 XP_012814707.1 5 - 6 586 71
XM_012959260.2 XP_012814714.1 6 - 7 976 71
GenBank mRNA BC075116.1 - 4 - 5 514 71

View Full Product Details