Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rwdd1
Entrez Gene ID
448293
Gene Name
RWD domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 69288313 - 69302610 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005815.1 NP_001005815.1 1 - 2 160 104
GenBank mRNA BC075365.1 - 1 - 2 160 104
CR761667.2 - 1 - 2 132 104

View Full Product Details