Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
eif6
Entrez Gene ID
448711
Gene Name
eukaryotic translation initiation factor 6
Gene Aliases
b(2)gcn, cab, eif3a, itgb4bp, p27(BBP), p27BBP
Chromosome Location
Chr.10: 6202626 - 6213577 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006883.1 NP_001006884.1 1 - 2 224 92
GenBank mRNA BC077665.1 - 1 - 2 224 92
CR760524.2 - 1 - 2 187 92
CR848359.2 - 2 - 3 278 92

View Full Product Details