Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
surf6
Entrez Gene ID
493251
Gene Name
surfeit 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683446: 244580 - 255216 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007864.1 NP_001007865.1 3 - 4 365 93
XM_012968261.2 XP_012823715.1 3 - 4 379 93
GenBank mRNA BC079967.1 - 3 - 4 365 93
CR926263.1 - 3 - 4 383 93
CR942359.2 - 2 - 3 363 93

View Full Product Details