Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rwdd4
Entrez Gene ID
493313
Gene Name
RWD domain containing 4
Gene Aliases
fam28a, rwdd4a
Chromosome Location
Chr.1: 35862765 - 35872882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007934.1 NP_001007935.1 2 - 3 247 82
GenBank mRNA BC080441.1 - 2 - 3 247 82

View Full Product Details