Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stx5
Entrez Gene ID
493353
Gene Name
syntaxin 5
Gene Aliases
stx5a
Chromosome Location
Chr.4: 41246214 - 41257215 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007990.1 NP_001007991.1 10 - 11 956 71
XM_012953584.2 XP_012809038.1 10 - 11 1044 71
XM_012953585.2 XP_012809039.1 10 - 11 1055 71
GenBank mRNA BC080503.1 - 10 - 11 956 71
CR926422.2 - 9 - 10 896 71

View Full Product Details