Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
m6pr
Entrez Gene ID
493477
Gene Name
mannose-6-phosphate receptor (cation dependent)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683450: 742370 - 755354 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008114.1 NP_001008115.1 3 - 4 565 92
XM_012956817.2 XP_012812271.1 4 - 5 543 92
XM_012956818.2 XP_012812272.1 3 - 4 435 92
GenBank mRNA BC081319.1 - 3 - 4 565 92

View Full Product Details