Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fam53a
Entrez Gene ID
493500
Gene Name
family with sequence similarity 53 member A
Gene Aliases
dntnp
Chromosome Location
Chr.1: 13456417 - 13506476 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008137.1 NP_001008138.1 3 - 4 446 70
XM_012960486.2 XP_012815940.1 3 - 4 421 70
XM_012960496.2 XP_012815950.1 3 - 4 609 70
GenBank mRNA BC081349.1 - 3 - 4 446 70

View Full Product Details