Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ywhae
Entrez Gene ID
493518
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
Gene Aliases
14-3-3e, 14-3-3epsilon, kcip-1, mdcr, mds
Chromosome Location
Chr.2: 39501294 - 39524429 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008155.1 NP_001008156.1 2 - 3 437 99
XM_012956823.1 XP_012812277.1 2 - 3 504 99
GenBank mRNA BC081369.1 - 2 - 3 437 99
CR848241.2 - 2 - 3 297 99

View Full Product Details