Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fgf8
Entrez Gene ID
493525
Gene Name
fibroblast growth factor 8
Gene Aliases
FGF-8, Xfgf-8, Xfgf8, aigf, fgf-8b, fgf8a, fgf8b, hbgf-8
Chromosome Location
Chr.7: 22701083 - 22710915 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008162.1 NP_001008163.1 2 - 3 378 94
GenBank mRNA BC082344.1 - 2 - 3 378 94

View Full Product Details