Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prss27
Entrez Gene ID
496619
Gene Name
serine protease 27
Gene Aliases
Xesp-1, caph2, marapsin, mpn, pancreasin, tmprss9
Chromosome Location
Chr.9: 2798854 - 2812905 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018097584.1 XP_017953073.1 5 - 6 930 69
GenBank mRNA BC087611.1 - 5 - 6 693 69

View Full Product Details