Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ass1
Entrez Gene ID
496645
Gene Name
argininosuccinate synthase 1
Gene Aliases
ass
Chromosome Location
Chr.8: 5543534 - 5589627 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011212.1 NP_001011212.1 13 - 14 1257 64
GenBank mRNA BC087767.1 - 13 - 14 1257 64

View Full Product Details