Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bhmt
Entrez Gene ID
496651
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016684775: 119 - 9739 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011217.1 NP_001011217.1 3 - 4 286 64
GenBank mRNA BC087774.1 - 3 - 4 286 64
BC089235.1 - 3 - 4 270 64

View Full Product Details