Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtg2
Entrez Gene ID
496677
Gene Name
mitochondrial ribosome-associated GTPase 2
Gene Aliases
gtpbp5
Chromosome Location
Chr.10: 1157211 - 1161391 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011236.1 NP_001011236.1 2 - 3 384 76
XM_012971705.1 XP_012827159.1 3 - 4 561 76
XM_012971707.2 XP_012827161.1 3 - 4 479 76
XM_012971708.1 XP_012827162.1 3 - 4 644 76
XM_012971709.2 XP_012827163.1 3 - 4 518 76
GenBank mRNA BC087807.1 - 2 - 3 384 76
CR942701.1 - 2 - 3 416 76

View Full Product Details