Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vdac3
Entrez Gene ID
496695
Gene Name
voltage-dependent anion channel 3
Gene Aliases
hd-vdac3
Chromosome Location
Chr.3: 120265942 - 120272269 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011249.2 NP_001011249.1 6 - 7 707 96
XM_012958925.2 XP_012814379.1 8 - 9 806 96
XM_012958926.2 XP_012814380.1 7 - 8 803 96
XM_012958927.2 XP_012814381.1 8 - 9 730 96
XM_012958929.1 XP_012814383.1 8 - 9 786 96
XM_012958930.2 XP_012814384.1 8 - 9 792 96
XM_012958931.1 XP_012814385.1 7 - 8 783 96
XM_012958932.2 XP_012814386.1 7 - 8 787 96
GenBank mRNA BC087828.1 - 6 - 7 708 96

View Full Product Details