Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agpat4
Entrez Gene ID
496716
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-agpat4, lpaat-delta
Chromosome Location
Chr.5: 42713661 - 42753277 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011265.1 NP_001011265.1 - 1208 98
GenBank mRNA BC087973.1 - - 1208 98

View Full Product Details