Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chtop
Entrez Gene ID
496726
Gene Name
chromatin target of PRMT1
Gene Aliases
c1orf77, fop, pp7704, srag
Chromosome Location
Chr.Un|NW_016684574: 6738 - 11753 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011274.1 NP_001011274.1 - 186 91
GenBank mRNA BC087986.1 - - 186 91

View Full Product Details