Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnase1l3
Entrez Gene ID
394515
Gene Name
deoxyribonuclease I-like 3
Gene Aliases
dhp2, dnas1l3
Chromosome Location
Chr.Un|NW_016683423: 422902 - 429459 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203588.1 NP_988919.1 6 - 7 725 91
GenBank mRNA BC061621.1 - 6 - 7 725 91
BC080151.1 - 6 - 7 91

View Full Product Details