Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cxorf56
Entrez Gene ID
394969
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.8: 21033865 - 21101640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204012.2 NP_989343.1 2 - 3 276 77
XM_018096190.1 XP_017951679.1 2 - 3 332 77
GenBank mRNA BC064195.1 - 2 - 3 257 77
BC084906.1 - 2 - 3 276 77
CR760470.2 - 2 - 3 257 77

View Full Product Details