Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
xgb
Entrez Gene ID
496620
Gene Name
x globin
Gene Aliases
gbX
Chromosome Location
Chr.8: 93897899 - 93906007 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011196.1 NP_001011196.1 1 - 2 175 100
GenBank mRNA AJ634915.1 - 1 - 2 175 100

View Full Product Details