Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
impact
Entrez Gene ID
496763
Gene Name
impact RWD domain protein
Gene Aliases
Ximpact, impact-A
Chromosome Location
Chr.6: 95881737 - 95895506 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011308.1 NP_001011308.1 5 - 6 562 115
GenBank mRNA BC088032.1 - 5 - 6 562 115
CR760285.2 - 5 - 6 569 115

View Full Product Details